Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
frm-10
Entrez Gene ID
179828
Gene Name
protein-coding
Gene Aliases
CELE_F25H9.5
Chromosome Location
Chr.V: 13463128 - 13467131 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001269578.3 NP_001256507.1 2 - 3 249 93

View Full Product Details