Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
eea-1
Entrez Gene ID
179832
Gene Name
protein-coding
Gene Aliases
CELE_T10G3.5
Chromosome Location
Chr.V: 13491067 - 13497372 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028956.4 NP_001024127.1 8 - 9 2946 78
NM_001028957.5 NP_001024128.1 8 - 9 2940 78
GenBank mRNA AY032860.1 - 8 - 9 2941 78

View Full Product Details