Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
bir-2
Entrez Gene ID
179841
Gene Name
Baculoviral IAP repeat-containing protein bir-2
Gene Aliases
CELE_C50B8.2
Chromosome Location
Chr.V: 13568846 - 13570030 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073961.5 NP_506362.1 1 - 2 154 105
GenBank mRNA U72208.1 - 1 - 2 150 105

View Full Product Details