The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_073985.6 | NP_506386.1 | 4 - 5 | 532 | 112 |
GenBank mRNA | AF536544.1 | - | 4 - 5 | 532 | 112 |