Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ceh-24
Entrez Gene ID
179877
Gene Name
Homeobox protein ceh-24
Gene Aliases
CELE_F55B12.1
Chromosome Location
Chr.V: 13813231 - 13815350 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074018.5 NP_506419.3 5 - 6 660 68

View Full Product Details