Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lbp-7
Entrez Gene ID
191701
Gene Name
Fatty acid-binding protein homolog 7
Gene Aliases
CELE_T22G5.2
Chromosome Location
Chr.V: 13879552 - 13880213 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074039.2 NP_506440.1 1 - 2 210 105

View Full Product Details