Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lbp-8
Entrez Gene ID
188761
Gene Name
FABP domain-containing protein
Gene Aliases
CELE_T22G5.6
Chromosome Location
Chr.V: 13890679 - 13891364 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074043.5 NP_506444.2 1 - 1 239 78

View Full Product Details