Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
srh-24
Entrez Gene ID
187767
Gene Name
Serpentine Receptor, class H
Gene Aliases
CELE_R10D12.11
Chromosome Location
Chr.V: 13958746 - 13961016 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074065.2 NP_506466.2 1 - 2 150 115

View Full Product Details