Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
end-3
Entrez Gene ID
191631
Gene Name
GATA-type domain-containing protein
Gene Aliases
CELE_F58E10.5
Chromosome Location
Chr.V: 14014695 - 14015980 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074079.3 NP_506480.1 1 - 2 261 104

View Full Product Details