Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ocam-1
Entrez Gene ID
187978
Gene Name
ONECUT (homeobox) Associated Motif containing
Gene Aliases
CELE_T02B5.2
Chromosome Location
Chr.V: 14167105 - 14167729 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074106.4 NP_506507.2 1 - 2 91 99

View Full Product Details