Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ent-3
Entrez Gene ID
179926
Gene Name
Equilibrative Nucleoside Transporter
Gene Aliases
CELE_K02E11.1
Chromosome Location
Chr.V: 14232281 - 14236076 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074120.4 NP_506521.2 5 - 6 696 103

View Full Product Details