Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nxf-1
Entrez Gene ID
191736
Gene Name
Nuclear RNA export factor 1
Gene Aliases
CELE_C15H11.3
Chromosome Location
Chr.V: 14411636 - 14415014 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136408.3 NP_001129880.1 2 - 3 528 76

View Full Product Details