Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
oig-3
Entrez Gene ID
190105
Gene Name
Ig-like domain-containing protein
Gene Aliases
CELE_Y50E8A.3
Chromosome Location
Chr.V: 14728954 - 14731134 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074236.4 NP_506637.1 4 - 5 354 86

View Full Product Details