Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hil-1
Entrez Gene ID
179993
Gene Name
Histone H1.X
Gene Aliases
CELE_C30G7.1
Chromosome Location
Chr.V: 14949777 - 14952800 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074279.6 NP_506680.1 3 - 4 321 76
GenBank mRNA AF002679.1 - 3 - 4 294 76

View Full Product Details