Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nmr-2
Entrez Gene ID
180002
Gene Name
protein-coding
Gene Aliases
CELE_T01C3.10
Chromosome Location
Chr.V: 15006341 - 15011660 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001269711.3 NP_001256640.1 11 - 12 1987 118
NM_001269712.1 NP_001256641.1 2 - 3 280 118
GenBank mRNA AF318614.2 - 11 - 12 1963 118

View Full Product Details