Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
unc-76
Entrez Gene ID
180014
Gene Name
protein-coding
Gene Aliases
CELE_C01G10.11
Chromosome Location
Chr.V: 15071577 - 15080152 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001269716.3 NP_001256645.1 1 - 2 207 75
NM_074311.6 NP_506712.4 1 - 2 390 75
NM_171553.5 NP_741641.2 1 - 2 204 75

View Full Product Details