Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srr-7
Entrez Gene ID
187957
Gene Name
Serpentine Receptor, class R
Gene Aliases
CELE_T01G5.4
Chromosome Location
Chr.V: 15128255 - 15129986 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074330.2 NP_506731.2 3 - 4 488 142

View Full Product Details