Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cyn-7
Entrez Gene ID
180027
Gene Name
Peptidyl-prolyl cis-trans isomerase 7
Gene Aliases
CELE_Y75B12B.2
Chromosome Location
Chr.V: 15185900 - 15187378 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074348.10 NP_506749.1 1 - 2 88 69

View Full Product Details