Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srw-15
Entrez Gene ID
183380
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_C41G6.3
Chromosome Location
Chr.V: 15230543 - 15231955 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074365.2 NP_506766.1 1 - 2 605 100

View Full Product Details