Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srx-51
Entrez Gene ID
188398
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_T10H4.9
Chromosome Location
Chr.V: 15281899 - 15283562 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074385.4 NP_506786.2 7 - 8 909 110

View Full Product Details