Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rpl-38
Entrez Gene ID
180044
Gene Name
60S ribosomal protein L38
Gene Aliases
CELE_C06B8.8
Chromosome Location
Chr.V: 15512179 - 15512775 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074459.7 NP_506860.1 1 - 2 155 60

View Full Product Details