Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
glb-14
Entrez Gene ID
184751
Gene Name
GLOBIN domain-containing protein
Gene Aliases
CELE_F21A3.6
Chromosome Location
Chr.V: 15543106 - 15544994 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074466.4 NP_506867.2 3 - 4 555 83

View Full Product Details