Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- atx-3
- Entrez Gene ID
-
180049
- Gene Name
- Ataxin-3 homolog
- Gene Aliases
-
CELE_F28F8.6
- Chromosome Location
-
Chr.V: 15575502 - 15577445 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001383468.1 |
|
5 - 6 |
821 |
85 |
- Gene Symbol
- mdt-28
- Entrez Gene ID
-
180048
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_F28F8.5
- Chromosome Location
-
Chr.V: 15574098 - 15576890 on Build WBcel235
View Full Product Details