Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sru-42
Entrez Gene ID
183829
Gene Name
Serpentine Receptor, class U
Gene Aliases
CELE_C55A1.8
Chromosome Location
Chr.V: 15646822 - 15652531 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074493.3 NP_506894.2 5 - 6 2469 136

View Full Product Details