Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nhr-51
Entrez Gene ID
180052
Gene Name
Nuclear hormone receptor family member nhr-51
Gene Aliases
CELE_K06B4.1
Chromosome Location
Chr.V: 15674007 - 15676003 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074497.5 NP_506898.2 2 - 3 414 86

View Full Product Details