Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nhr-52
Entrez Gene ID
3565623
Gene Name
Nuclear hormone receptor family member nhr-52
Gene Aliases
CELE_K06B4.2
Chromosome Location
Chr.V: 15677522 - 15680074 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074498.4 NP_506899.1 3 - 4 820 109

View Full Product Details