Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
str-38
Entrez Gene ID
188783
Gene Name
Seven TM Receptor
Gene Aliases
CELE_T23D5.2
Chromosome Location
Chr.V: 15729202 - 15732328 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074515.7 NP_506916.3 2 - 3 629 77

View Full Product Details