Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
str-18
Entrez Gene ID
188785
Gene Name
Seven TM Receptor
Gene Aliases
CELE_T23D5.6
Chromosome Location
Chr.V: 15740370 - 15741951 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074518.3 NP_506919.2 2 - 3 565 80

View Full Product Details