Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nhr-60
Entrez Gene ID
180059
Gene Name
Nuclear hormone receptor family member nhr-60
Gene Aliases
CELE_F57A10.5
Chromosome Location
Chr.V: 15770268 - 15773241 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074529.4 NP_506930.1 3 - 4 666 73

View Full Product Details