Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srbc-55
Entrez Gene ID
3564908
Gene Name
Serpentine Receptor, class BC (class B-like)
Gene Aliases
CELE_F54B8.16
Chromosome Location
Chr.V: 15829022 - 15830007 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028794.1 NP_001023965.1 1 - 2 175 156

View Full Product Details