Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
oac-42
Entrez Gene ID
188167
Gene Name
protein-coding
Gene Aliases
CELE_T06C12.8
Chromosome Location
Chr.V: 15881932 - 15884839 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276972.3 NP_001263901.1 6 - 7 1014 139
NM_001276973.2 NP_001263902.1 6 - 7 1063 139

View Full Product Details