Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cgt-1
Entrez Gene ID
188169
Gene Name
Ceramide glucosyltransferase 1
Gene Aliases
CELE_T06C12.10
Chromosome Location
Chr.V: 15886716 - 15888851 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074570.5 NP_506971.2 1 - 2 51 73

View Full Product Details