Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srj-10
Entrez Gene ID
184488
Gene Name
Serpentine Receptor, class J
Gene Aliases
CELE_F14H3.1
Chromosome Location
Chr.V: 16072765 - 16073956 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074645.1 NP_507046.1 2 - 3 355 108

View Full Product Details