Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
str-25
Entrez Gene ID
185759
Gene Name
Seven TM Receptor
Gene Aliases
CELE_F44G3.11
Chromosome Location
Chr.V: 16115681 - 16116897 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001322602.1 NP_001309465.1 3 - 4 963 103

View Full Product Details