Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
str-99
Entrez Gene ID
184276
Gene Name
Seven TM Receptor
Gene Aliases
CELE_F10A3.6
Chromosome Location
Chr.V: 16157335 - 16158491 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074669.2 NP_507070.2 2 - 3 279 76

View Full Product Details