Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gcy-20
Entrez Gene ID
184797
Gene Name
Receptor-type guanylate cyclase gcy-20
Gene Aliases
CELE_F21H7.9
Chromosome Location
Chr.V: 16248583 - 16253787 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074700.1 NP_507101.1 8 - 9 1504 60

View Full Product Details