Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srxa-7
Entrez Gene ID
183073
Gene Name
Serpentine Receptor, class XA
Gene Aliases
CELE_C31A11.6
Chromosome Location
Chr.V: 16307553 - 16309688 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074718.2 NP_507119.1 3 - 4 911 98

View Full Product Details