Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srbc-65
Entrez Gene ID
189567
Gene Name
Serpentine Receptor, class BC (class B-like)
Gene Aliases
CELE_Y32B12B.3
Chromosome Location
Chr.V: 16550980 - 16552282 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074788.1 NP_507189.1 1 - 2 175 123

View Full Product Details