Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
oac-53
Entrez Gene ID
189254
Gene Name
NRF domain-containing protein
Gene Aliases
CELE_W06G6.1
Chromosome Location
Chr.V: 16623857 - 16627970 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074804.3 NP_507205.2 10 - 11 1937 89

View Full Product Details