Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srz-99
Entrez Gene ID
3564956
Gene Name
Serpentine Receptor, class Z
Gene Aliases
CELE_T25E12.13
Chromosome Location
Chr.V: 16730261 - 16731464 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028985.1 NP_001024156.1 1 - 2 147 149

View Full Product Details