Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dkf-2
Entrez Gene ID
180121
Gene Name
protein-coding
Gene Aliases
CELE_T25E12.4
Chromosome Location
Chr.V: 16747559 - 16788435 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001129550.2 NP_001123022.1 5 - 6 973 109
NM_001269795.3 NP_001256724.1 10 - 11 1508 109
NM_001269796.3 NP_001256725.1 10 - 11 1514 109
NM_001269797.3 NP_001256726.1 6 - 7 848 109
NM_074838.6 NP_507239.2 10 - 11 1577 109

View Full Product Details