Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dkf-2
Entrez Gene ID
180121
Gene Name
protein-coding
Gene Aliases
CELE_T25E12.4
Chromosome Location
Chr.V: 16747559 - 16788435 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001129550.2 NP_001123022.1 4 - 5 867 94
NM_001269796.3 NP_001256725.1 9 - 10 1408 94
NM_001269797.3 NP_001256726.1 5 - 6 742 94
NM_074838.6 NP_507239.2 9 - 10 1471 94

View Full Product Details