Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
clec-31
Entrez Gene ID
186002
Gene Name
C-type LECtin
Gene Aliases
CELE_F49A5.2
Chromosome Location
Chr.V: 16856419 - 16858077 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074857.3 NP_507258.2 3 - 4 431 101

View Full Product Details