Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srd-68
Entrez Gene ID
184234
Gene Name
Serpentine Receptor, class D (delta)
Gene Aliases
CELE_F09C6.7
Chromosome Location
Chr.V: 16901215 - 16902826 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074870.2 NP_507271.2 4 - 5 718 59

View Full Product Details