Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fbxa-90
Entrez Gene ID
190842
Gene Name
F-box domain-containing protein
Gene Aliases
CELE_Y102A5C.19
Chromosome Location
Chr.V: 16966814 - 16968586 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074889.5 NP_507290.1 1 - 2 175 94

View Full Product Details