Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srz-54
Entrez Gene ID
190848
Gene Name
Serpentine Receptor, class Z
Gene Aliases
CELE_Y102A5C.25
Chromosome Location
Chr.V: 16980668 - 16981880 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074895.1 NP_507296.1 2 - 3 278 114

View Full Product Details