Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srbc-14
Entrez Gene ID
188135
Gene Name
Serpentine Receptor, class BC (class B-like)
Gene Aliases
CELE_T05E12.2
Chromosome Location
Chr.V: 17070527 - 17071678 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074921.2 NP_507322.2 2 - 3 346 133

View Full Product Details