Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srz-47
Entrez Gene ID
190523
Gene Name
Serpentine Receptor, class Z
Gene Aliases
CELE_Y68A4A.2
Chromosome Location
Chr.V: 17190831 - 17192339 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074953.3 NP_507354.2 4 - 5 732 90

View Full Product Details