Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srz-48
Entrez Gene ID
184729
Gene Name
Serpentine Receptor, class Z
Gene Aliases
CELE_F20E11.1
Chromosome Location
Chr.V: 17458533 - 17460161 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_075018.1 NP_507419.1 4 - 5 739 107

View Full Product Details