Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srw-72
Entrez Gene ID
184734
Gene Name
Serpentine Receptor, class W
Gene Aliases
CELE_F20E11.6
Chromosome Location
Chr.V: 17467051 - 17468561 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001322603.1 NP_001309457.1 3 - 4 746 144

View Full Product Details